Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-CSF Antibody (67604), Biotin, Novus Biologicals™
SDP

Catalog No. NB370816 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370816 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370816 Supplier Novus Biologicals Supplier No. FAB414B
Add to Cart
Add to Cart

Rat Monoclonal Antibody

G-CSF Monoclonal specifically detects G-CSF in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Neutralization, Western Blot.

Specifications

Antigen G-CSF
Applications ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Neutralization, Western Blot
Classification Monoclonal
Clone 67604
Conjugate Biotin
Dilution Western Blot, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Formulation PBS
Gene Alias C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3OS, filgrastim, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin
Host Species Rat
Immunogen E. coli-derived recombinant mouse G-CSF, Val31-Ala208, Accession # P09920
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 1440
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.