Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-CSF Antibody (CSF3/3166R), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB380966 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB380966 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB380966 Supplier Novus Biologicals Supplier No. NBP308381AF647
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

G-CSF Monoclonal antibody specifically detects G-CSF in Human, Macaque samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen G-CSF
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CSF3/3166R
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3OS, filgrastim, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin
Host Species Rabbit
Immunogen Recombinant full-length human G-CSF protein (Uniprot: P09919)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 1440
Target Species Human, Macaque
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.