Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ G-CSF Antibody Pair [HRP]
SDP

Catalog No. p-7183320 Shop All R&D Systems Products
Click to view available options
Quantity:
15 Plates

Monoclonal/Polyclonal Antibody Pair

This solid phase sandwich ELISA utilizes an antibody specific for Human G-CSF.

Specifications

Assay Range 93.75 pg/mL
Assay Sensitivity 93.75-6000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Human G-CSF.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3OS, filgrastim, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin
Gene ID (Entrez) 1440
Gene Symbol CSF3
Host Species Rabbit
Kit Contents
  • Standard: Each vial contains 305 ng of recombinant Human G-CSF.
  • Capture Antibody: 1 mg/mL of rabbit anti-Human G-CSF polyclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human G-CSF monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research
    Storage Requirements Storage is content dependent.
    Target G-CSF
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 6000 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.