Learn More
G-CSF Rabbit anti-Human, Macaque, Clone: CSF3/3166R, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | G-CSF |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CSF3/3166R |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3OS, filgrastim, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.