Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-CSFR/CD114 Antibody (38643), PE, Novus Biologicals™
SDP

Catalog No. NB296366 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB296366 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB296366 Supplier Novus Biologicals Supplier No. FAB3811P
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

G-CSFR/CD114 Monoclonal specifically detects G-CSFR/CD114 in Human samples. It is validated for CyTOF, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Flow Cytometry, Western Blot.

Specifications

Antigen G-CSFR/CD114
Applications CyTOF, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Flow Cytometry, Western Blot
Classification Monoclonal
Clone 38643
Conjugate PE
Dilution Western Blot, Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF-ready
Formulation PBS
Gene Alias CD114, CD114 antigen, colony stimulating factor 3 receptor (granulocyte), G-CSF receptor, GCSFRG-CSF-R, granulocyte colony-stimulating factor receptor
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human G-CSF R/CD114, Glu25-Pro621, Accession # Q99062
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1441
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.