Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G gamma7 Antibody (OTI3G1), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB270838A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB270838A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB270838A48 Supplier Novus Biologicals Supplier No. NBP270838AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

G gamma7 Monoclonal specifically detects G gamma7 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen G gamma7
Applications Western Blot
Classification Monoclonal
Clone OTI3G1
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ00058, GNGT7, guanine nucleotide binding protein (G protein), gamma 7, guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-7
Gene Symbols GNG7
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 2-65 of human G gamma7 (NP_443079) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2788.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.