Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G protein alpha 12 Antibody, Novus Biologicals™
SDP

Catalog No. NBP155321 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP155321 100 μL
NB15743020 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP155321 Supplier Novus Biologicals Supplier No. NBP155321
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

G protein alpha 12 Polyclonal specifically detects G protein alpha 12 in Human samples. It is validated for Western Blot.

Specifications

Antigen G protein alpha 12
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q03113
Gene Alias G alpha-12, gep, G-protein subunit alpha-12, guanine nucleotide binding protein (G protein) alpha 12, guanine nucleotide-binding protein subunit alpha-12, MGC104623, MGC99644, NNX3, RMP, WUGSC:H_GS165O14.2
Gene Symbols GNA12
Host Species Rabbit
Immunogen Synthetic peptides corresponding to GNA12(guanine nucleotide binding protein (G protein) alpha 12) The peptide sequence was selected from the middle region of GNA12. Peptide sequence TFQLYVPALSALWRDSGIREAFSRRSEFQLGESVKYFLDNLDRIGQLNYF.
Molecular Weight of Antigen 44 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2768
Test Specificity This product is specific to Subunit or Isoform: alpha-12.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.