Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G protein alpha inhibitor 1 Antibody (7H7), Novus Biologicals™
SDP

Catalog No. NBP229768 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229768 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP229768 Supplier Novus Biologicals Supplier No. NBP229768

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

G protein alpha inhibitor 1 Monoclonal specifically detects G protein alpha inhibitor 1 in Human samples. It is validated for Western Blot.

Specifications

Antigen G protein alpha inhibitor 1
Applications Western Blot
Classification Monoclonal
Clone 7H7
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Gene Accession No. P63096
Gene Alias Adenylate cyclase-inhibiting G alpha protein, Gi, Gi1 protein alpha subunit, guanine nucleotide binding protein (G protein), alpha inhibiting activitypolypeptide 1, guanine nucleotide-binding protein G(i) subunit alpha-1
Gene Symbols GNAI1
Host Species Mouse
Immunogen Peptide corresponding to residues 110-123 of GiAlpha 1.
Molecular Weight of Antigen 41 kDa
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2770
Test Specificity Vertebrates.
Target Species Other
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.