Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G protein alpha inhibitor 1 Antibody (7H7), Novus Biologicals™

Mouse Monoclonal Antibody
$631.00
Specifications
Antigen | G protein alpha inhibitor 1 |
---|---|
Clone | 7H7 |
Dilution | Western Blot 1:100-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
G protein alpha inhibitor 1 Monoclonal specifically detects G protein alpha inhibitor 1 in Human samples. It is validated for Western Blot.Specifications
G protein alpha inhibitor 1 | |
Western Blot 1:100-1:2000 | |
Monoclonal | |
Mouse | |
Signal Transduction | |
P63096 | |
2770 | |
Peptide corresponding to residues 110-123 of GiAlpha 1. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
41 kDa |
7H7 | |
Western Blot | |
Unconjugated | |
RUO | |
Other | |
Adenylate cyclase-inhibiting G alpha protein, Gi, Gi1 protein alpha subunit, guanine nucleotide binding protein (G protein), alpha inhibiting activitypolypeptide 1, guanine nucleotide-binding protein G(i) subunit alpha-1 | |
GNAI1 | |
IgG2b | |
Affinity Purified | |
Vertebrates. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title