Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G protein beta 4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179681
Description
G protein beta 4 Polyclonal specifically detects G protein beta 4 in Human samples. It is validated for Western Blot.Specifications
G protein beta 4 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
G protein beta-4 subunit, guanine nucleotide binding protein (G protein), beta polypeptide 4, guanine nucleotide binding protein beta subunit 4, guanine nucleotide-binding protein subunit beta-4, Transducin beta chain 4 | |
Rabbit | |
37 kDa | |
100 μL | |
Signal Transduction | |
59345 | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
NP_067642 | |
GNB4 | |
Synthetic peptide directed towards the middle region of human GNB4. Peptide sequence DGMCRQSFTGHVSDINAVSFFPNGYAFATGSDDATCRLFDLRADQELLLY. | |
Affinity purified | |
RUO | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction