Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G protein beta 4 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen G protein beta 4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP179681
SDP
View Documents
Novus Biologicals
NBP179681
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

G protein beta 4 Polyclonal specifically detects G protein beta 4 in Human samples. It is validated for Western Blot.
Specifications

Specifications

G protein beta 4
Polyclonal
Rabbit
Signal Transduction
G protein beta-4 subunit, guanine nucleotide binding protein (G protein), beta polypeptide 4, guanine nucleotide binding protein beta subunit 4, guanine nucleotide-binding protein subunit beta-4, Transducin beta chain 4
GNB4
IgG
37 kDa
Western Blot
Unconjugated
RUO
NP_067642
59345
Synthetic peptide directed towards the middle region of human GNB4. Peptide sequence DGMCRQSFTGHVSDINAVSFFPNGYAFATGSDDATCRLFDLRADQELLLY.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.