Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-substrate Antibody (OTI2A8), PerCP, Novus Biologicals™
SDP

Catalog No. NB288735 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288735 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288735 Supplier Novus Biologicals Supplier No. NBP271645PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

G-substrate Monoclonal antibody specifically detects G-substrate in Human samples. It is validated for Western Blot

Specifications

Antigen G-substrate
Applications Western Blot
Classification Monoclonal
Clone OTI2A8
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias chromosome 7 open reading frame 16, GSBSG-substrate
Host Species Mouse
Immunogen Full length human recombinant protein of human C7ORF16 (NP_006649) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10842
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.