Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-substrate Antibody (OTI2E5), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP271644V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271644V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271644V Supplier Novus Biologicals Supplier No. NBP271644V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

G-substrate Monoclonal specifically detects G-substrate in Human samples. It is validated for Western Blot.

Specifications

Antigen G-substrate
Applications Western Blot
Classification Monoclonal
Clone OTI2E5
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias chromosome 7 open reading frame 16, GSBSG-substrate
Gene Symbols PPP1R17
Host Species Mouse
Immunogen Full length recombinant protein of human G-substrate (NP_006649) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10842.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.