Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G2A/GPR132 Rabbit anti-Human, Mouse, Monkey, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB019785 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019785 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB019785 Supplier Novus Biologicals Supplier No. NBP2690900.025ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

G2A/GPR132 Polyclonal specifically detects G2A/GPR132 in Human, Mouse, Monkey samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen G2A/GPR132
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation PBS containing 50% glycerol, 0.5% BSA
Gene Alias G protein-coupled receptor 132, G2 accumulation protein, G2AG protein-coupled receptor G2A, MGC99642, probable G-protein coupled receptor 132
Host Species Rabbit
Immunogen Synthesized peptide derived from the C-terminal region of human G2A.
Purification Method Affinity purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 29933
Target Species Human, Mouse, Monkey
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.