Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G2A/GPR132 Rabbit anti-Human, Mouse, Monkey, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$207.00
Specifications
Antigen | G2A/GPR132 |
---|---|
Applications | Western Blot, ELISA, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
G2A/GPR132 Polyclonal specifically detects G2A/GPR132 in Human, Mouse, Monkey samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
G2A/GPR132 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Monkey | |
G protein-coupled receptor 132, G2 accumulation protein, G2AG protein-coupled receptor G2A, MGC99642, probable G-protein coupled receptor 132 | |
Synthesized peptide derived from the C-terminal region of human G2A. | |
Primary | |
Store at -20°C. |
Western Blot, ELISA, Immunofluorescence | |
Unconjugated | |
Rabbit | |
GPCR | |
PBS containing 50% glycerol, 0.5% BSA | |
29933 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title