Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G3BP2 Antibody, Novus Biologicals™
SDP

Catalog No. NB433250 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB433250 25 μL
NBP182976 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB433250 Supplier Novus Biologicals Supplier No. NBP18297625UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

G3BP2 Polyclonal antibody specifically detects G3BP2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen G3BP2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias G3BP-2, GAP SH3 domain-binding protein 2, GTPase activating protein (SH3 domain) binding protein 2, KIAA0660, ras GTPase-activating protein-binding protein 2, Ras-GTPase activating protein SH3 domain-binding protein 2
Gene Symbols G3BP2
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:KNLEELEEKSTTPPPAEPVSLPQEPPKPRVEAKPEVQSQPPRVREQRPRERPGFPPRGPRPGRGDMEQNDS
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9908
Test Specificity Specificity of human sG3BP2 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.