Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ G5

Catalog No. p-7166820
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Ubiquitin isopeptidase inhibitor; induces apoptosis

G5 is an ubiquitin isopeptidase inhibitor; activates caspases and apoptosis in an apoptosome-independent pathway.

Specifications

Quantity 10 mg
Molecular Formula C19H14N2O7S
Formula Weight 414.39
Product Type Inhibitor
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.