Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G5pr Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP189907
Description
G5pr Polyclonal specifically detects G5pr in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
G5pr | |
Polyclonal | |
Western Blot 0.04 - 0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
Q969Q6 | |
PPP2R3C | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LLDIENKGYLNVFSLNYFFRAIQELMKIHGQDPVSFQDVKDEIFDMVKPKDPLKISLQDLINSNQGDTVTTILIDLNGFWT | |
Affinity Purified | |
RUO | |
55012 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
C14orf10, chromosome 14 open reading frame 10, FLJ20644, G4-1, G5pr, protein phosphatase 2 (formerly 2A), regulatory subunit B'', gamma, protein phosphatase 2, regulatory subunit B'', gamma, Protein phosphatase subunit G5PR, rhabdomyosarcoma antigen MU-RMS-40.6A, Rhabdomyosarcoma antigen MU-RMS-40.6A/6C, rhabdomyosarcoma antigen Mu-RMS-40.6C, serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit gamma | |
Rabbit | |
53 kDa | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction