Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G6b/C6orf25 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094037 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094037 100 μL
NB094038 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094037 Supplier Novus Biologicals Supplier No. NBP303530100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

G6b/C6orf25 Polyclonal specifically detects G6b/C6orf25 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen G6b/C6orf25
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias chromosome 6 open reading frame 25, G6b, immunoglobulin receptor, MGC142279, MGC142281, NG31, protein G6b
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 18-142 of human G6b/C6orf25 (NP_612121.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80739
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.