Learn More
Description
Specifications
Specifications
| Antigen | G6PC2 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA 1:10000, Immunohistochemistry, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:50 |
| Formulation | Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide |
| Gene Alias | EC 3.1.3.9, G6Pase 2, G-6-Pase 2, glucose-6-phosphatase 2, glucose-6-phosphatase, catalytic, 2, IGRPMGC141936, islet-specific G6CP-related protein, Islet-specific glucose-6-phosphatase catalytic subunit-related protein, islet-specific glucose-6-phosphatase-related protein |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide taken within amino acid region 80-130 on G6PC2 protein, and common in larger variant of glucose 6 phosphatase. |
| Purification Method | Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
