Learn More
Blackline Safety™ G7x Satellite-Connected Safety Monitor with Multi-Gas Pump Sensor Cartridge, North America
Shop All Blackline Safety Corporation ProductsDescription
- Fall detection and No motion detection: Tri-axis accelerometer, tri-axis gyro, software processing, configurable sensitivity, configurable time window (1 to 30 mins, or off ) for no motion detection
- Emergency latch: Pull latch to trigger remote alert
- Silent emergency: Press and hold latch to trigger remote alert
- Low-battery: configurable threshold
- Worker check-in: Configurable timer (5 to 180 min, or off ), automatic check-in when driving
Specifications
Specifications
| Description | G7x 900MHz |
| Components Measured | Hydrogen Sulfide, LEL-I, Carbon Monoxide, Oxygen |
| Type | Multigas Cartridge |