Learn More
Blackline Safety™ G7c Cellular-Connected Safety Monitor with Multi-Gas Pump Sensor Cartridge, North America
Wireless connectivity ensures that each G7c is remotely configurable and firmware updates are automatic. G7c supports three cartridge types that provide a choice of gas detection options. Everything is completely customizable to meet the needs of every industry, application and scenario
| Catalog Number | Mfr. No. | Components Measured | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Components Measured | Price | Quantity & Availability | |||||
|
17-000-0001
|
Blackline Safety™
G7CPDOPVNA |
Carbon Monoxide/Hydrogen Sulfide Twin Tox, Oxygen, LEL-P, PID | N/A | N/A | N/A | ||||
|
17-000-0070
|
Blackline Safety™
G7CPHIMONA |
Hydrogen Sulfide, LEL-I, Carbon Monoxide, Oxygen | N/A | N/A | N/A | ||||
|
17-000-0071
|
Blackline Safety™
G7CPHIXXNA |
Hydrogen Sulfide, LEL-I | N/A | N/A | N/A | ||||
|
17-000-0073
|
Blackline Safety™
G7CPHMOPNA |
Hydrogen Sulfide, Carbon Monoxide, Oxygen, LEL-P | N/A | N/A | N/A | ||||
Description
- Fall detection and No motion detection: Tri-axis accelerometer, tri-axis gyro, software processing, configurable sensitivity, configurable time window (1 to 30 mins, or off ) for no motion detection
- Emergency latch: Pull latch to trigger remote alert
- Silent emergency: Press and hold latch to trigger remote alert
- Low-battery: configurable threshold
- Worker check-in: Configurable timer (5 to180 min, or off ), automatic check-in when driving
Specifications
| Multigas Cartridge |