Learn More
Blackline Safety™ G7c Cellular-Connected Safety Monitor with Multi-Gas Sensor Cartridge, North America
Catalog No. 170000046 Shop All Blackline Safety Corporation Products
Change view
Click to view available options
Components Measured:
Carbon Dioxide, Hydrogen Sulfide, LEL-I, Oxygen
Carbon Dioxide, LEL-I, Carbon Monoxide, Oxygen
Carbon Dioxide, LEL-I, Oxygen
Carbon Dioxide, Oxygen
Carbon Dioxide, Oxygen, LEL-P
Carbon Monoxide, Oxygen
Carbon Monoxide, Oxygen, LEL-P
Carbon Monoxide, Oxygen, LEL-P, PID
Chlorine, Carbon Monoxide, Oxygen, LEL-P
Chlorine, Oxygen, LEL-P, Cobalt Hydride
Hydrogen Sulfide, Carbon Monoxide
Hydrogen Sulfide, Carbon Monoxide, Oxygen
Hydrogen Sulfide, Carbon Monoxide, Oxygen, LEL-P
Hydrogen Sulfide, LEL-I, Chlorine, Oxygen
Hydrogen Sulfide, LEL-I, Oxygen
Hydrogen Sulfide, LEL-I, Oxygen, Carbon Monoxide - H
Hydrogen Sulfide, LEL-I, Oxygen, PID
Hydrogen Sulfide, LEL-I, Oxygen, Sulfur Dioxide
Hydrogen Sulfide, Oxygen, Carbon Monoxide - H
Hydrogen Sulfide, Oxygen, LEL-P
Hydrogen Sulfide, Oxygen, LEL-P, Carbon Monoxide - H
LEL-I, Carbon Monoxide, Oxygen
LEL-I, Carbon Monoxide, Oxygen, Sulfur Dioxide
LEL-I, Chlorine, Carbon Monoxide, Oxygen
LEL-I, Chlorine, Oxygen
LEL-I, Chlorine, Oxygen, Sulfur Dioxide
LEL-I, Oxygen
LEL-I, Oxygen, PID
LEL-I, PID
Methane, Carbon Dioxide
Methane, Carbon Dioxide, LEL-I, Oxygen
Methane, Carbon Dioxide, Oxygen
Methane, Carbon Dioxide, Oxygen, LEL-P
Methane, Carbon Monoxide
Methane, Carbon Monoxide, Oxygen, LEL-P
Methane, Hydrogen Sulfide, LEL-I, Oxygen
Methane, Hydrogen Sulfide, Oxygen
Methane, LEL-I, Carbon Monoxide, Oxygen
Methane, LEL-I, Chlorine, Oxygen
Methane, LEL-I, Oxygen
Methane, LEL-I, Oxygen, Cobalt Hydride
Methane, LEL-I, Oxygen, Sulfur Dioxide
Methane, Oxygen
Oxygen, LEL-P
Oxygen, LEL-P, Cobalt Hydride, PID
Oxygen, LEL-P, PID
Oxygen, PID
48 product options available for selection
Product selection table with 48 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-000-0046 LEL-I, Carbon Monoxide, Oxygen, Sulfur Dioxide
17-000-0002 Methane, Carbon Dioxide, LEL-I, Oxygen
17-000-0003 Methane, Carbon Dioxide, Oxygen, LEL-P
17-000-0004 Methane, Carbon Dioxide, Oxygen
17-000-0005 Methane, Carbon Dioxide
17-000-0008 Methane, Hydrogen Sulfide, LEL-I, Oxygen
17-000-0009 Methane, Hydrogen Sulfide, Oxygen
17-000-0010 Methane, LEL-I, Chlorine, Oxygen
17-000-0011 Methane, LEL-I, Carbon Monoxide, Oxygen
17-000-0012 Methane, LEL-I, Oxygen, Cobalt Hydride
17-000-0013 Methane, LEL-I, Oxygen, Sulfur Dioxide
17-000-0014 Methane, LEL-I, Oxygen
17-000-0015 Methane, Carbon Monoxide, Oxygen, LEL-P
17-000-0016 Methane, Carbon Monoxide
17-000-0017 Methane, Oxygen
17-000-0020 Carbon Dioxide, Hydrogen Sulfide, LEL-I, Oxygen
17-000-0021 Carbon Dioxide, LEL-I, Carbon Monoxide, Oxygen
17-000-0022 Carbon Dioxide, LEL-I, Oxygen
17-000-0024 Carbon Dioxide, Oxygen, LEL-P
17-000-0025 Carbon Dioxide, Oxygen
17-000-0031 Hydrogen Sulfide, LEL-I, Chlorine, Oxygen
17-000-0033 Hydrogen Sulfide, LEL-I, Oxygen, Carbon Monoxide - H
17-000-0034 Hydrogen Sulfide, LEL-I, Oxygen, Sulfur Dioxide
17-000-0035 Hydrogen Sulfide, LEL-I, Oxygen, PID
17-000-0036 Hydrogen Sulfide, LEL-I, Oxygen
17-000-0037 Hydrogen Sulfide, Carbon Monoxide, Oxygen, LEL-P
17-000-0038 Hydrogen Sulfide, Carbon Monoxide, Oxygen
17-000-0039 Hydrogen Sulfide, Carbon Monoxide
17-000-0040 Hydrogen Sulfide, Oxygen, LEL-P, Carbon Monoxide - H
17-000-0041 Hydrogen Sulfide, Oxygen, LEL-P
17-000-0042 Hydrogen Sulfide, Oxygen, Carbon Monoxide - H
17-000-0043 LEL-I, Chlorine, Carbon Monoxide, Oxygen
17-000-0044 LEL-I, Chlorine, Oxygen, Sulfur Dioxide
17-000-0045 LEL-I, Chlorine, Oxygen
17-000-0047 LEL-I, Carbon Monoxide, Oxygen
17-000-0048 LEL-I, Oxygen, PID
17-000-0049 LEL-I, Oxygen
17-000-0050 LEL-I, PID
17-000-0051 Chlorine, Carbon Monoxide, Oxygen, LEL-P
17-000-0052 Chlorine, Oxygen, LEL-P, Cobalt Hydride
17-000-0053 Carbon Monoxide, Oxygen, LEL-P, PID
17-000-0054 Carbon Monoxide, Oxygen, LEL-P
17-000-0055 Carbon Monoxide, Oxygen
17-000-0056 Oxygen, LEL-P, Cobalt Hydride, PID
17-000-0057 Oxygen, LEL-P, PID
17-000-0058 Oxygen, LEL-P
17-000-0059 Oxygen, PID
17-000-0023 Carbon Dioxide, Oxygen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 48 options available.
48 options
Catalog No. 17-000-0046 Supplier Blackline Safety™ Supplier No. G7CQIMOSNA
Wireless connectivity ensures that each G7c is remotely configurable and firmware updates are automatic. G7c supports three cartridge types that provide a choice of gas detection options. Everything is completely customizable to meet the needs of every industry, application and scenario
  • Fall detection and No motion detection: Tri-axis accelerometer, tri-axis gyro, software processing, configurable sensitivity, configurable time window (1 to 30 mins, or off ) for no motion detection
  • Emergency latch: Pull latch to trigger remote alert
  • Silent emergency: Press and hold latch to trigger remote alert
  • Low-battery: configurable threshold
  • Worker check-in: Configurable timer (5 to180 min, or off ), automatic check-in when driving
Specifications
Components Measured LEL-I, Carbon Monoxide, Oxygen, Sulfur Dioxide
Type Multigas Cartridge