Learn More
Blackline Safety™ G7x Satellite-Connected Safety Monitor with Multi-Gas/Twin Tox Sensor Cartridge, North America
Catalog No. 170000100 Shop All Blackline Safety Corporation Products
Change view
Click to view available options
Components Measured:
Carbon Dioxide, Carbon Monoxide/Hydrogen Sulfide Twin Tox, LEL-I, Oxygen
Carbon Dioxide, Carbon Monoxide/Hydrogen Sulfide Twin Tox, Oxygen, LEL-P
Carbon Monoxide/Hydrogen Sulfide Twin Tox, LEL-I, Oxygen, PID
Carbon Monoxide/Hydrogen Sulfide Twin Tox, LEL-I, Oxygen, Sulfur Dioxide
Carbon Monoxide/Hydrogen Sulfide Twin Tox, Oxygen, LEL-P, PID
Carbon Monoxide/Hydrogen Sulfide Twin Tox, Oxygen, LEL-P, Sulfur Dioxide
Carbon Monoxide/Hydrogen Sulfide Twin Tox, PID
Methane, Carbon Monoxide/Hydrogen Sulfide Twin Tox, LEL-I, Oxygen
Methane, Carbon Monoxide/Hydrogen Sulfide Twin Tox, Oxygen, LEL-P
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-000-0100 Carbon Monoxide/Hydrogen Sulfide Twin Tox, LEL-I, Oxygen, PID
17-000-0079 Methane, Carbon Monoxide/Hydrogen Sulfide Twin Tox, LEL-I, Oxygen
17-000-0080 Methane, Carbon Monoxide/Hydrogen Sulfide Twin Tox, Oxygen, LEL-P
17-000-0091 Carbon Dioxide, Carbon Monoxide/Hydrogen Sulfide Twin Tox, LEL-I, Oxygen
17-000-0092 Carbon Dioxide, Carbon Monoxide/Hydrogen Sulfide Twin Tox, Oxygen, LEL-P
17-000-0099 Carbon Monoxide/Hydrogen Sulfide Twin Tox, LEL-I, Oxygen, Sulfur Dioxide
17-000-0101 Carbon Monoxide/Hydrogen Sulfide Twin Tox, Oxygen, LEL-P, Sulfur Dioxide
17-000-0102 Carbon Monoxide/Hydrogen Sulfide Twin Tox, Oxygen, LEL-P, PID
17-000-0103 Carbon Monoxide/Hydrogen Sulfide Twin Tox, PID
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17-000-0100 Supplier Blackline Safety™ Supplier No. G7XQDIOVNA
Wireless connectivity ensures that each G7x is remotely configurable and firmware updates are automatic. G7x supports three cartridge types that provide a choice of gas detection options. Everything is completely customizable to meet the needs of every industry, application and scenario
  • Fall detection and No motion detection: Tri-axis accelerometer, tri-axis gyro, software processing, configurable sensitivity, configurable time window (1 to 30 mins, or off ) for no motion detection
  • Emergency latch: Pull latch to trigger remote alert
  • Silent emergency: Press and hold latch to trigger remote alert
  • Low-battery: configurable threshold
  • Worker check-in: Configurable timer (5 to 180 min, or off ), automatic check-in when driving
Specifications
Components Measured Carbon Monoxide/Hydrogen Sulfide Twin Tox, LEL-I, Oxygen, PID
Type Multigas Cartridge
Frequency 900 MHz