Learn More
Blackline Safety™ G7x Satellite-Connected Safety Monitor with Multi-Gas Sensor Cartridge, North America
Catalog No. 170000118 Shop All Blackline Safety Corporation Products
Change view
Click to view available options
Components Measured:
Carbon Dioxide, Hydrogen Sulfide, LEL-I, Oxygen
Carbon Dioxide, LEL-I, Carbon Monoxide, Oxygen
Carbon Dioxide, LEL-I, Oxygen
Carbon Dioxide, Oxygen
Carbon Dioxide, Oxygen, LEL-P
Carbon Monoxide, Oxygen
Carbon Monoxide, Oxygen, LEL-P
Carbon Monoxide, Oxygen, LEL-P, PID
Chlorine, Carbon Monoxide, Oxygen, LEL-P
Chlorine, Oxygen, LEL-P, Carbon Monoxide - H
Hydrogen Sulfide, Carbon Monoxide
Hydrogen Sulfide, Carbon Monoxide, Oxygen
Hydrogen Sulfide, Carbon Monoxide, Oxygen, LEL-P
Hydrogen Sulfide, LEL-I, Carbon Monoxide, Oxygen
Hydrogen Sulfide, LEL-I, Chlorine, Oxygen
Hydrogen Sulfide, LEL-I, Oxygen
Hydrogen Sulfide, LEL-I, Oxygen, Carbon Monoxide - H
Hydrogen Sulfide, LEL-I, Oxygen, PID
Hydrogen Sulfide, LEL-I, Oxygen, Sulfur Dioxide
Hydrogen Sulfide, Oxygen, Carbon Monoxide - H
Hydrogen Sulfide, Oxygen, LEL-P
Hydrogen Sulfide, Oxygen, LEL-P, Carbon Monoxide - H
LEL-I, Carbon Monoxide, Oxygen
LEL-I, Carbon Monoxide, Oxygen, Sulfur Dioxide
LEL-I, Chlorine, Carbon Monoxide, Oxygen
LEL-I, Chlorine, Oxygen
LEL-I, Chlorine, Oxygen, Sulfur Dioxide
LEL-I, Oxygen
LEL-I, Oxygen, PID
LEL-I, PID
Methane, Carbon Dioxide
Methane, Carbon Dioxide, LEL-I, Oxygen
Methane, Carbon Dioxide, Oxygen
Methane, Carbon Dioxide, Oxygen, LEL-P
Methane, Carbon Monoxide
Methane, Carbon Monoxide, Oxygen, LEL-P
Methane, Hydrogen Sulfide, LEL-I, Oxygen
Methane, Hydrogen Sulfide, Oxygen
Methane, LEL-I, Carbon Monoxide, Oxygen
Methane, LEL-I, Chlorine, Oxygen
Methane, LEL-I, Oxygen
Methane, LEL-I, Oxygen, Carbon Monoxide - H
Methane, LEL-I, Oxygen, Sulfur Dioxide
Methane, Oxygen
Oxygen, LEL-P
Oxygen, LEL-P, Carbon Monoxide - H, PID
Oxygen, LEL-P, PID
Oxygen, PID
49 product options available for selection
Product selection table with 49 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-000-0118 LEL-I, Chlorine, Oxygen
17-000-0075 Methane, Carbon Dioxide, LEL-I, Oxygen
17-000-0076 Methane, Carbon Dioxide, Oxygen, LEL-P
17-000-0077 Methane, Carbon Dioxide, Oxygen
17-000-0078 Methane, Carbon Dioxide
17-000-0081 Methane, Hydrogen Sulfide, LEL-I, Oxygen
17-000-0082 Methane, Hydrogen Sulfide, Oxygen
17-000-0083 Methane, LEL-I, Chlorine, Oxygen
17-000-0084 Methane, LEL-I, Carbon Monoxide, Oxygen
17-000-0085 Methane, LEL-I, Oxygen, Carbon Monoxide - H
17-000-0086 Methane, LEL-I, Oxygen, Sulfur Dioxide
17-000-0087 Methane, LEL-I, Oxygen
17-000-0088 Methane, Carbon Monoxide, Oxygen, LEL-P
17-000-0089 Methane, Carbon Monoxide
17-000-0090 Methane, Oxygen
17-000-0093 Carbon Dioxide, Hydrogen Sulfide, LEL-I, Oxygen
17-000-0094 Carbon Dioxide, LEL-I, Carbon Monoxide, Oxygen
17-000-0095 Carbon Dioxide, LEL-I, Oxygen
17-000-0096 Carbon Dioxide, Oxygen
17-000-0097 Carbon Dioxide, Oxygen, LEL-P
17-000-0098 Carbon Dioxide, Oxygen
17-000-0104 Hydrogen Sulfide, LEL-I, Chlorine, Oxygen
17-000-0105 Hydrogen Sulfide, LEL-I, Carbon Monoxide, Oxygen
17-000-0106 Hydrogen Sulfide, LEL-I, Oxygen, Carbon Monoxide - H
17-000-0107 Hydrogen Sulfide, LEL-I, Oxygen, Sulfur Dioxide
17-000-0108 Hydrogen Sulfide, LEL-I, Oxygen, PID
17-000-0109 Hydrogen Sulfide, LEL-I, Oxygen
17-000-0110 Hydrogen Sulfide, Carbon Monoxide, Oxygen, LEL-P
17-000-0111 Hydrogen Sulfide, Carbon Monoxide, Oxygen
17-000-0112 Hydrogen Sulfide, Carbon Monoxide
17-000-0113 Hydrogen Sulfide, Oxygen, LEL-P, Carbon Monoxide - H
17-000-0114 Hydrogen Sulfide, Oxygen, LEL-P
17-000-0115 Hydrogen Sulfide, Oxygen, Carbon Monoxide - H
17-000-0116 LEL-I, Chlorine, Carbon Monoxide, Oxygen
17-000-0117 LEL-I, Chlorine, Oxygen, Sulfur Dioxide
17-000-0119 LEL-I, Carbon Monoxide, Oxygen, Sulfur Dioxide
17-000-0120 LEL-I, Carbon Monoxide, Oxygen
17-000-0121 LEL-I, Oxygen, PID
17-000-0122 LEL-I, Oxygen
17-000-0123 LEL-I, PID
17-000-0124 Chlorine, Carbon Monoxide, Oxygen, LEL-P
17-000-0125 Chlorine, Oxygen, LEL-P, Carbon Monoxide - H
17-000-0126 Carbon Monoxide, Oxygen, LEL-P, PID
17-000-0127 Carbon Monoxide, Oxygen, LEL-P
17-000-0128 Carbon Monoxide, Oxygen
17-000-0129 Oxygen, LEL-P, Carbon Monoxide - H, PID
17-000-0130 Oxygen, LEL-P, PID
17-000-0131 Oxygen, LEL-P
17-000-0132 Oxygen, PID
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 49 options available.
49 options
Catalog No. 17-000-0118 Supplier Blackline Safety™ Supplier No. G7XQILOXNA
Wireless connectivity ensures that each G7x is remotely configurable and firmware updates are automatic. G7x supports three cartridge types that provide a choice of gas detection options. Everything is completely customizable to meet the needs of every industry, application and scenario
  • Fall detection and No motion detection: Tri-axis accelerometer, tri-axis gyro, software processing, configurable sensitivity, configurable time window (1 to 30 mins, or off ) for no motion detection
  • Emergency latch: Pull latch to trigger remote alert
  • Silent emergency: Press and hold latch to trigger remote alert
  • Low-battery: configurable threshold
  • Worker check-in: Configurable timer (5 to 180 min, or off ), automatic check-in when driving
Specifications
Components Measured LEL-I, Chlorine, Oxygen
Type Multigas Cartridge
Frequency 900 MHz