Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G9a/EHMT2 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB292058 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292058 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292058 Supplier Novus Biologicals Supplier No. NB10040825APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

G9a/EHMT2 Polyclonal antibody specifically detects G9a/EHMT2 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot (Negative)

Specifications

Antigen G9a/EHMT2
Applications Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative)
Formulation PBS
Gene Alias ankyrin repeat-containing protein, BAT8, C6orf30, chromosome 6 open reading frame 30, DKFZp686H08213, EC 2.1.1.-, EC 2.1.1.43, euchromatic histone-lysine N-methyltransferase 2HLA-B-associated transcript 8, FLJ35547, G9A histone methyltransferase, G9AEm:AF134726.3, H3-K9-HMTase 3, Histone H3-K9 methyltransferase 3, histone-lysine N-methyltransferase, H3 lysine-9 specific 3, HLA-B associated transcript 8, KMT1CNG36/G9a, Lysine N-methyltransferase 1C, NG36, Protein G9a
Host Species Rabbit
Immunogen Maps to a region between residue 1160 and the C-terminus (residue 1210) of human G9A (Euchromatic Histone-Lysine N-methyltransferase 2) using the numbering given in entry NP_006700.2 (GeneID 10919).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10919
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.