Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G9a/EHMT2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP242846 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242846 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242846 Supplier Novus Biologicals Supplier No. NBP242846
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects G9a/EHMT2 in Mouse samples, and it is validated for Western Blotting

G9a/EHMT2 Polyclonal specifically detects G9a/EHMT2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen G9a/EHMT2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Alias ankyrin repeat-containing protein, BAT8, C6orf30, chromosome 6 open reading frame 30, DKFZp686H08213, EC 2.1.1.-, EC 2.1.1.43, euchromatic histone-lysine N-methyltransferase 2HLA-B-associated transcript 8, FLJ35547, G9A histone methyltransferase, G9AEm:AF134726.3, H3-K9-HMTase 3, Histone H3-K9 methyltransferase 3, histone-lysine N-methyltransferase, H3 lysine-9 specific 3, HLA-B associated transcript 8, KMT1CNG36/G9a, Lysine N-methyltransferase 1C, NG36, Protein G9a
Gene Symbols EHMT2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of mouse G9a. The exact sequence is proprietary.
Molecular Weight of Antigen 138 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10919
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.