Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAB1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$343.50 - $573.00

Specifications

Antigen GAB1
Dilution Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB159463
SDP
View Documents
Novus Biologicals
NBP31771625UL
25 μg
Each for $343.50
Only null left
Add to Cart
 
NB159462
SDP
View Documents
Novus Biologicals
NBP317716100UL
100 μg
Each for $573.00
Only null left
Add to Cart
 
Description

Description

GAB1 Polyclonal antibody specifically detects GAB1 in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

GAB1
Western Blot, Immunofluorescence
Unconjugated
Rabbit
Apoptosis, Signal Transduction, Tyrosine Kinases
PBS, pH 7.2, 40% glycerol
2549
IgG
Affinity purified
Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
Polyclonal
Purified
RUO
Human
GRB2-associated binder 1, GRB2-associated binding protein 1, GRB2-associated-binding protein 1, Growth factor receptor bound protein 2-associated protein 1
This antibody was developed against Recombinant Protein corresponding to amino acids: TSKLDTIPDIPPPRPPKPHPAHDRSPVETCSIPRTASDTDSSYCIPTAGMSPSRSNTISTVDLNKLRKDASSQDCYDIPRAFP
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.