Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAB2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP216564 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP216564 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP216564 Supplier Novus Biologicals Supplier No. NBP216564
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GAB2 Polyclonal antibody specifically detects GAB2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen GAB2
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9UQC2
Gene Alias GRB2-associated binding protein 2, GRB2-associated-binding protein 2, Growth factor receptor bound protein 2-associated protein 2, KIAA0571GRB2-associated binder 2, pp100
Gene Symbols GAB2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GAB2. The exact sequence is proprietary.
Molecular Weight of Antigen 74 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 9846
Test Specificity Human. Expected to cross react based on sequence identity: Mouse (91%), Rat (90%).
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.