Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA-A R beta 3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP106988 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106988 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP106988 Supplier Novus Biologicals Supplier No. NBP106988
Add to Cart
Add to Cart

Goat Polyclonal Antibody

GABA-A R beta 3 Polyclonal specifically detects GABA-A R beta 3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen GABA-A R beta 3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 2 - 4 ug/ml, Immunohistochemistry-Paraffin 2 - 4 ug/ml, Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_000805.1
Gene Alias ECA5, GABA(A) receptor beta-3 subunit, GABA(A) receptor subunit beta-3, GABAA receptor beta-3 subunit, GABA-alpha receptor beta-2 subunit, gamma-aminobutyric acid (GABA) A receptor, beta 3, gamma-aminobutyric acid receptor subunit beta-3, MGC9051
Gene Symbols GABRB3
Host Species Goat
Immunogen Peptide with sequence C-EKTAKAKNDRSK corresponding to internal region according to NP_068712.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Neuronal Cell Markers, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 2562
Test Specificity This antibody is expected to recognize both reported isoforms (NP_000805.1, NP_068712.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.