Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA-A R delta Antibody (N151/3), Biotin, Novus Biologicals™
SDP

Catalog No. NBP147616B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147616B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP147616B Supplier Novus Biologicals Supplier No. NBP147616B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GABA-A R delta Monoclonal specifically detects GABA-A R delta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GABA-A R delta
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S151-3
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P18506
Gene Alias EIG10, EJM7, GABA(A) receptor subunit delta, GABA-A receptor, delta polypeptide, gamma-aminobutyric acid (GABA) A receptor, delta, gamma-aminobutyric acid receptor subunit delta, GEFSP5, MGC45284
Gene Symbols GABRD
Host Species Mouse
Immunogen Synthetic peptide amino acids 15-34 of rat GABA-A-R-Delta
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2563
Test Specificity Detects approx 55kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.