Learn More
Description
Specifications
Specifications
| Antigen | GABA-A R delta |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | S151-3 |
| Conjugate | HRP |
| Dilution | Western Blot, Immunohistochemistry |
| Formulation | PBS with No Preservative |
| Gene Accession No. | P18506 |
| Gene Alias | EIG10, EJM7, GABA(A) receptor subunit delta, GABA-A receptor, delta polypeptide, gamma-aminobutyric acid (GABA) A receptor, delta, gamma-aminobutyric acid receptor subunit delta, GEFSP5, MGC45284 |
| Gene Symbols | GABRD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
