Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA-A R gamma 2 Antibody, Novus Biologicals™
SDP

Catalog No. NB300151 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300151 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300151 Supplier Novus Biologicals Supplier No. NB300151
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 6 publications

GABA-A R gamma 2 Polyclonal specifically detects GABA-A R gamma 2 in Human, Mouse, Rat, Turkey, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated.

Specifications

Antigen GABA-A R gamma 2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:400, Immunohistochemistry-Frozen
Formulation Neat whole antisera with No Preservative
Gene Accession No. P22723, P18508
Gene Alias CAE2, ECA2, GABA(A) receptor subunit gamma-2, gamma-aminobutyric acid (GABA) A receptor, gamma 2, gamma-aminobutyric acid receptor subunit gamma-2, GEFSP3
Gene Symbols GABRG2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acid residues specific to the gamma 2 subunit conjugated to KLH. Accession # P18508
Molecular Weight of Antigen 46 kDa
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 2566
Test Specificity Specific for the ∼46k gamma2-subunit of the GABAA receptor in Western blots of rat brain extracts.
Target Species Human, Mouse, Rat, Turkey, Zebrafish
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.