Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABA-A R gamma 3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB10056662
Description
GABA-A R gamma 3 Polyclonal specifically detects GABA-A R gamma 3 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
GABA-A R gamma 3 | |
Polyclonal | |
Western Blot 1:250-1:1000 | |
Q99928 | |
GABRG3 | |
A synthetic peptide corresponding to amino acids somewhere between 10-60 of human GABAA receptor. | |
Unpurified | |
RUO | |
Primary | |
In rat brain, a 51 kDa band is observed. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
Whole antisera with PBS with 0.05% Sodium Azide | |
GABA(A) receptor subunit gamma-3, gamma-aminobutyric acid (GABA) A receptor, gamma 3, gamma-aminobutyric acid receptor subunit gamma-3 | |
Rabbit | |
54 kDa | |
0.2 mL | |
Signal Transduction | |
2567 | |
Human, Mouse, Rat | |
Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction