Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABA-A R gamma 3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$206.50 - $426.00
Specifications
Antigen | GABA-A R gamma 3 |
---|---|
Dilution | Western Blot 1:250-1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056662T
![]() |
Novus Biologicals
NB10056662SS |
0.05 mL |
Each for $206.50
|
|
|||||
NB10056662
![]() |
Novus Biologicals
NB10056662 |
0.2 mL |
Each for $426.00
|
|
|||||
Description
GABA-A R gamma 3 Polyclonal specifically detects GABA-A R gamma 3 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
GABA-A R gamma 3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
Whole antisera with PBS with 0.05% Sodium Azide | |
GABA(A) receptor subunit gamma-3, gamma-aminobutyric acid (GABA) A receptor, gamma 3, gamma-aminobutyric acid receptor subunit gamma-3 | |
GABRG3 | |
IgG | |
Unpurified | |
In rat brain, a 51 kDa band is observed. |
Western Blot 1:250-1:1000 | |
Polyclonal | |
Antisera | |
RUO | |
Human, Mouse, Rat | |
Q99928 | |
2567 | |
A synthetic peptide corresponding to amino acids somewhere between 10-60 of human GABAA receptor. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
54 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title