Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA-A R gamma 3 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 2 publications

Supplier:  Novus Biologicals NB10056662SS

 View more versions of this product

Catalog No. NB10056662T


Only null left
Add to Cart

Description

Description

GABA-A R gamma 3 Polyclonal specifically detects GABA-A R gamma 3 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

GABA-A R gamma 3
Polyclonal
Western Blot 1:250-1:1000
Q99928
GABRG3
A synthetic peptide corresponding to amino acids somewhere between 10-60 of human GABAA receptor.
Unpurified
RUO
Primary
In rat brain, a 51 kDa band is observed.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
Whole antisera with PBS with 0.05% Sodium Azide
GABA(A) receptor subunit gamma-3, gamma-aminobutyric acid (GABA) A receptor, gamma 3, gamma-aminobutyric acid receptor subunit gamma-3
Rabbit
54 kDa
0.05 mL
Signal Transduction
2567
Human, Mouse, Rat
Antisera
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only