Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA-A R rho 1 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094762 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094762 100 μL
NB094763 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094762 Supplier Novus Biologicals Supplier No. NBP303524100UL

Rabbit Polyclonal Antibody

GABA-A R rho 1 Polyclonal specifically detects GABA-A R rho 1 in Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen GABA-A R rho 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias GABA(C) receptor, gamma-aminobutyric acid (GABA) A receptor, rho-1, gamma-aminobutyric acid (GABA) receptor, rho 1, gamma-aminobutyric acid receptor subunit rho-1, rho 1)
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-80 of human GABA-A R rho 1 (NP_002033.2).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2569
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.