Learn More
Description
Specifically detects GABA in All samples, and it is validated for Dot Blot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | GABA |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:100-1:1000, Dot Blot |
| Formulation | 0.1M Tris-Glycine (pH7), 20% Glycerol with 0.01% Thimerosal |
| Gene Alias | y-Aminobutyric acid |
| Gene Symbols | GABA |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
