Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA-AR alpha 3 Antibody, Novus Biologicals™
SDP

Catalog No. NB10061096 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10061096 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10061096 Supplier Novus Biologicals Supplier No. NB10061096
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

GABA-AR alpha 3 Polyclonal specifically detects GABA-AR alpha 3 in Mouse, Rat samples. It is validated for Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen GABA-AR alpha 3
Applications Gene Knock-Out, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:100
Formulation 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
Gene Accession No. P20236
Gene Alias GABA(A) receptor subunit alpha-3, gamma-aminobutyric acid (GABA) A receptor, alpha 3, gamma-aminobutyric acid receptor subunit alpha-3, gamma-animobutyric acid (GABA) A receptor, alpha 3, MGC33793
Gene Symbols GABRA3
Host Species Rabbit
Immunogen Synthetic peptide from the N-terminal region of the alpha 3 subunit. Accession # P20236
Molecular Weight of Antigen 51 kDa
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2556
Test Specificity Specific for the ∼51k alpha 3-subunit of the GABAA receptor in Western blots. Labeling is absent in alpha 3-subunit knockout animals.
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.