Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA-AR alpha 3 Antibody, Novus Biologicals™
SDP

Catalog No. NB158098 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB158098 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB158098 Supplier Novus Biologicals Supplier No. NBP314326
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GABA-AR alpha 3 Polyclonal antibody specifically detects GABA-AR alpha 3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen GABA-AR alpha 3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias GABA(A) receptor subunit alpha-3, gamma-aminobutyric acid (GABA) A receptor, alpha 3, gamma-aminobutyric acid receptor subunit alpha-3, gamma-animobutyric acid (GABA) A receptor, alpha 3, MGC33793
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from C-Terminus of human GABA-AR alpha 3.
Purification Method Immunogen affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2556
Target Species Human
Content And Storage Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Product Type Antibody
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.