Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA-AR alpha 4 Antibody (S398A-34), Novus Biologicals™
SDP

Catalog No. NBP259326 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259326 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259326 Supplier Novus Biologicals Supplier No. NBP259326
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GABA-AR alpha 4 Monoclonal specifically detects GABA-AR alpha 4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GABA-AR alpha 4
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S398A-34
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Formulation PBS pH7.4, 50% glycerol with 0.1% Sodium Azide
Gene Alias GABA(A) receptor subunit alpha-4, gamma-aminobutyric acid (GABA) A receptor, alpha 4, gamma-aminobutyric acid receptor subunit alpha-4
Gene Symbols GABRA4
Host Species Mouse
Immunogen Fusion protein amino acids 410-450 (Cytoplasmic C-terminus) of rat GABA-A-Receptor Alpha 4
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 2557
Test Specificity Detects 60kDa. Does not cross-react with other GABA-A-Receptor Alpha subunits.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.