Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA-AR alpha 5 Antibody (5C11), Novus Biologicals™
SDP

Catalog No. NB297855 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297855 100 μL
NB297854 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297855 Supplier Novus Biologicals Supplier No. NBP326561100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GABA-AR alpha 5 Monoclonal antibody specifically detects GABA-AR alpha 5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen GABA-AR alpha 5
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 5C11
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias GABA(A) receptor subunit alpha-5, gamma-aminobutyric acid (GABA) A receptor, alpha 5, gamma-aminobutyric acid receptor subunit alpha-5, MGC138184
Host Species Rabbit
Immunogen A synthesized peptide derived from Human GABA-AR alpha 5 [UniProt P31644]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cellular Markers, Neuronal Cell Markers, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 2558
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.