Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA-AR alpha 5 Antibody, Novus Biologicals™
SDP

Catalog No. NB300195 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300195 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300195 Supplier Novus Biologicals Supplier No. NB300195
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 5 publications

GABA-AR alpha 5 Polyclonal specifically detects GABA-AR alpha 5 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry Free-Floating.

Specifications

Antigen GABA-AR alpha 5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Free Floating)
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry, Immunohistochemistry Free-Floating
Formulation 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
Gene Accession No. P19969
Gene Alias GABA(A) receptor subunit alpha-5, gamma-aminobutyric acid (GABA) A receptor, alpha 5, gamma-aminobutyric acid receptor subunit alpha-5, MGC138184
Gene Symbols GABRA5
Host Species Rabbit
Immunogen Fusion protein from the cytoplasmic loop of the alpha 5 subunit of GABA-AR alpha 5. Accession # P19969
Molecular Weight of Antigen 55 kDa
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Neuronal Cell Markers, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 2558
Test Specificity Specific for endogenous levels of the ∼55k alpha 5-subunit of the GABAA receptor.
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.