Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA-AR alpha 5 Antibody (OTI1G9), PerCP, Novus Biologicals™
SDP

Catalog No. NB289380 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289380 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289380 Supplier Novus Biologicals Supplier No. NBP270789PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GABA-AR alpha 5 Monoclonal antibody specifically detects GABA-AR alpha 5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen GABA-AR alpha 5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI1G9
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias GABA(A) receptor subunit alpha-5, gamma-aminobutyric acid (GABA) A receptor, alpha 5, gamma-aminobutyric acid receptor subunit alpha-5, MGC138184
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 32-259 of human GABRA5(NP_000801) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Neuronal Cell Markers, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 2558
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.