Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA-AR alpha 6 Antibody, Novus Biologicals™
SDP

Catalog No. NB300150 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300150 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300150 Supplier Novus Biologicals Supplier No. NB300150
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GABA-AR alpha 6 Polyclonal specifically detects GABA-AR alpha 6 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen GABA-AR alpha 6
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:10-1:500
Formulation Neat whole antisera with No Preservative
Gene Accession No. P30191
Gene Alias GABA subunit A receptor alpha 6, GABA(A) receptor subunit alpha-6, gamma-aminobutyric acid (GABA) A receptor, alpha 6, gamma-aminobutyric acid receptor subunit alpha-6, MGC116903, MGC116904
Gene Symbols GABRA6
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acid residues specific to the alpha 6 subunit conjugated to KLH. Accession # P30191
Molecular Weight of Antigen 57 kDa
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Neuronal Cell Markers, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 2559
Test Specificity Labels the ∼57k alpha 6-subunit of the GABAA receptor in Western blots of rat brain extracts.
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.