Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA-B R1 Antibody (S93A-49), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP259336UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259336UV 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259336UV Supplier Novus Biologicals Supplier No. NBP259336UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GABA-B R1 Monoclonal specifically detects GABA-B R1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GABA-B R1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S93A-49
Conjugate DyLight 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ92613, GABABR1, GABBR1-3, gamma-aminobutyric acid (GABA) B receptor, 1, gamma-aminobutyric acid type B receptor subunit 1, GB1, GPRC3Asubunit 1c, hGB1a, seven transmembrane helix receptor
Gene Symbols GABBR1
Host Species Mouse
Immunogen Fusion protein amino acids 873-977 (cytoplasmic C-terminus) of rat GABA(B)R1
Purification Method Protein G purified
Quantity 100 μL
Research Discipline GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 2550
Test Specificity Detects 115kDa. No cross-reactivity against GABA(B)R2.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.