Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABA-B R2 Antibody (5G7B9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP237280
Description
GABA-B R2 Monoclonal specifically detects GABA-B R2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GABA-B R2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
G protein-coupled receptor 51, GABA-BR2, GABA-B-R2, GABABR2GABA-B receptor 2, gamma-aminobutyric acid (GABA) B receptor, 2, gamma-aminobutyric acid type B receptor subunit 2, Gb2, GPR51GABA-B receptor, R2 subunit, GPRC3BFLJ36928, G-protein coupled receptor 51, HG20HRIHFB2099 | |
Mouse | |
105.8 kDa | |
0.1 mL | |
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
9568 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
5G7B9 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
GABBR2 | |
Purified recombinant fragment of human GABBR2 (AA: 319-483) expressed in E. Coli. | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction