Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABA-B R2 Antibody (5G7B9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | GABA-B R2 |
---|---|
Clone | 5G7B9 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Description
GABA-B R2 Monoclonal specifically detects GABA-B R2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GABA-B R2 | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
G protein-coupled receptor 51, GABA-BR2, GABA-B-R2, GABABR2GABA-B receptor 2, gamma-aminobutyric acid (GABA) B receptor, 2, gamma-aminobutyric acid type B receptor subunit 2, Gb2, GPR51GABA-B receptor, R2 subunit, GPRC3BFLJ36928, G-protein coupled receptor 51, HG20HRIHFB2099 | |
GABBR2 | |
IgG2a | |
Affinity Purified |
5G7B9 | |
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
Monoclonal | |
Mouse | |
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
PBS with 0.05% Sodium Azide | |
9568 | |
Purified recombinant fragment of human GABBR2 (AA: 319-483) expressed in E. Coli. | |
Primary | |
105.8 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title