Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA-B R2 Antibody (S81-2), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB259335405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259335405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB259335405 Supplier Novus Biologicals Supplier No. NBP259335AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GABA-B R2 Monoclonal specifically detects GABA-B R2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GABA-B R2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S81-2
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias G protein-coupled receptor 51, GABA-BR2, GABA-B-R2, GABABR2GABA-B receptor 2, gamma-aminobutyric acid (GABA) B receptor, 2, gamma-aminobutyric acid type B receptor subunit 2, Gb2, GPR51GABA-B receptor, R2 subunit, GPRC3BFLJ36928, G-protein coupled receptor 51, HG20HRIHFB2099
Gene Symbols GABBR2
Host Species Mouse
Immunogen Fusion protein amino acids 861-912 of rat GABA(B)R2
Purification Method Protein G purified
Quantity 100 μL
Research Discipline GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 9568
Test Specificity Detects 105kDa. No cross-reactivity against GABA(B)R1.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.