Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA Transporter 2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP247147 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP247147 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP247147 Supplier Novus Biologicals Supplier No. NBP247147
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GABA Transporter 2 Polyclonal specifically detects GABA Transporter 2 in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen GABA Transporter 2
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:200 - 1:5000, ELISA, Immunohistochemistry 1:50 - 1:500
Formulation 0.1M Tris 0.1M Glycine and 2% Sucrose with No Preservative
Gene Accession No. P31646
Gene Alias GABA transport protein, GAT-2, GAT2Solute carrier family 6 member 13, sodium- and chloride-dependent GABA transporter 2, solute carrier family 6 (neurotransmitter transporter, GABA), member 13
Gene Symbols SLC6A13
Host Species Rabbit
Immunogen A 15 residue synthetic peptide derived from C-terminal domain of rat GAT-2.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6540
Test Specificity Reacts with GAT-2 from rat, mouse and human central nervous system.
Reconstitution Reconstitute with distilled water.
Target Species Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.